accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22951-96-8

IUPAC Name: (2S)-2-amino-3-(thiophen-2-yl)propanoic acid
Synonyms: β(2-thienyl)alanine
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: WTOFYLAWDLQMBZ-LURJTMIESA-N
SMILES: N[C@@H](CC1=CC=CS1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results