accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SHO

CAS RN 22962-49-8

IUPAC Name: (5-methyl-1-benzothiophen-2-yl)methanol
Synonyms: (5-methyl-1-benzothiophen-2-yl)methanol
Molecular Weight (g/mol): 178.25
Molecular Formula: C10H10OS
InChi Key: NPSSZIBANMLQHP-UHFFFAOYSA-N
SMILES: CC1=CC=C2SC(CO)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results