accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2304-94-1

IUPAC Name: 3-{[(benzyloxy)carbonyl]amino}propanoic acid
Synonyms: 3-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: GEVGRLPYQJTKKS-UHFFFAOYSA-N
SMILES: OC(=O)CCNC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)