accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2305-26-2

HOOOHO

CAS RN 2305-26-2

IUPAC Name: cyclohex-4-ene-1,2-dicarboxylic acid
Synonyms: cyclohex-4-ene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: ILUAAIDVFMVTAU-UHFFFAOYNA-N
SMILES: OC(=O)C1CC=CCC1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)