accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2307-10-0

IUPAC Name: 1-(propylsulfanyl)ethan-1-one
Synonyms: 1-(propylsulfanyl)ethanone
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: SBWFWBJCYMBZEY-UHFFFAOYSA-N
SMILES: CCCSC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results