accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23088-23-5

H3COONN

CAS RN 23088-23-5

IUPAC Name: methyl quinoxaline-6-carboxylate
Synonyms: methyl quinoxaline-6-carboxylate
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: VFPWJISMXACHIG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2N=CC=NC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results