accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23135-22-0

CH3HNOONSH3CONH3CH3C(Z)

CAS RN 23135-22-0

IUPAC Name: (Z)-[(dimethylcarbamoyl)(methylsulfanyl)methylidene]amino N-methylcarbamate
Synonyms: oxamyl
Molecular Weight (g/mol): 219.26
Molecular Formula: C7H13N3O3S
InChi Key: KZAUOCCYDRDERY-UITAMQMPSA-N
SMILES: CNC(=O)O\N=C(/SC)C(=O)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (5)
Solvent or Matrix 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)