accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClN

CAS RN 2315-81-3

IUPAC Name: 3-chloro-4-hydroxybenzonitrile
Synonyms: 3-chloro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: CRYPJUOSZDQWJZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results