accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBrN

CAS RN 2315-86-8

IUPAC Name: 3-bromo-4-hydroxybenzonitrile
Synonyms: 3-bromo-4-hydroxybenzonitrile
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: HLHNOIAOWQFNGW-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Br)C#N
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results