accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2316-26-9

IUPAC Name: (2E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
Synonyms: 3,4-dimethoxycinnamic acid cinnamic acid,3,4-dimethoxy
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: HJBWJAPEBGSQPR-GQCTYLIASA-N
SMILES: COC1=CC=C(\C=C\C(O)=O)C=C1OC
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)