accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFFFNCS

CAS RN 23165-29-9

IUPAC Name: 1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 271.18
Molecular Formula: C9H3F6NS
InChi Key: FXOSSGVJGGNASE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)