accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 231953-38-1

IUPAC Name: 3-fluoro-4-(trifluoromethyl)benzonitrile
Synonyms: 3-fluoro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: CFPNGJCBTZAVDN-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1C(F)(F)F)C#N
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (8)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)