accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)ON

CAS RN 23210-22-2

IUPAC Name: (3S)-3-methyl-1-phenyl-2,3-dihydro-1H-indol-2-one
Synonyms: (3S)-3-methyl-1-phenyl-3H-indol-2-one
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: DJKASMOURATDIE-NSHDSACASA-N
SMILES: C[C@@H]1C(=O)N(C2=CC=CC=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results