CAS RN 2324-98-3
IUPAC Name:
1,2,4,5-tetrafluoro-3-methoxybenzene
Synonyms:
2,3,5,6-tetrafluoroanisole
Molecular Weight (g/mol):
180.1
Molecular Formula:
C7H4F4O
InChi Key:
AXCOCGJDERQVDK-UHFFFAOYSA-N
SMILES:
COC1=C(F)C(F)=CC(F)=C1F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluoroanisole, 97%
CAS: 2324-98-3 Molecular Formula: C7H4F4O Molecular Weight (g/mol): 180.10 MDL Number: MFCD00000311 InChI Key: AXCOCGJDERQVDK-UHFFFAOYSA-N Synonym: 2,3,5,6-tetrafluoroanisole,2,3,5,6-tetrafluoromethoxy benzene,anisole, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluoromethoxybenzene,methyl 2,3,5,6-tetrafluorophenyl ether,benzene, 1,2,4,5-tetrafluoro-3-methoxy,1,2,4,5-tetrafluoro-3-methoxy-benzene,pubchem10114,acmc-1clho,2,3,5,6-tetrafluoro-anisole PubChem CID: 75351 IUPAC Name: 1,2,4,5-tetrafluoro-3-methoxybenzene SMILES: COC1=C(F)C(F)=CC(F)=C1F