accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23326-27-4

CH3OOH3C

CAS RN 23326-27-4

IUPAC Name: methyl but-2-ynoate
Synonyms: methyl but-2-ynoate
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: UJQCANQILFWSDJ-UHFFFAOYSA-N
SMILES: COC(=O)C#CC
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results