accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23351-07-7

IUPAC Name: 4-(1H-pyrrol-1-yl)benzonitrile
Synonyms: 4-(pyrrol-1-yl)benzonitrile
Molecular Weight (g/mol): 168.2
Molecular Formula: C11H8N2
InChi Key: OKVSZRKKRHNDOL-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results