accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23356-96-9

IUPAC Name: (pyrrolidin-2-yl)methanol
Synonyms: 2-pyrrolidinemethanol
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: HVVNJUAVDAZWCB-UHFFFAOYNA-N
SMILES: OCC1CCCN1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)