accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3N(R)(S)OH

CAS RN 23364-44-5

IUPAC Name: (1R,2S)-2-hydroxy-1,2-diphenylethan-1-aminium
Synonyms: (1R,2S)-2-hydroxy-1,2-diphenylethanaminium
Molecular Weight (g/mol): 214.29
Molecular Formula: C14H16NO
InChi Key: GEJJWYZZKKKSEV-KGLIPLIRSA-O
SMILES: [NH3+][C@@H]([C@@H](O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results