CAS RN 2338-75-2
IUPAC Name:
2-[4-(trifluoromethyl)phenyl]acetonitrile
Synonyms:
2-[4-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol):
185.15
Molecular Formula:
C9H6F3N
InChi Key:
QNKOCFJZJWOXDE-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(CC#N)C=C1
1
–
2
of
2
results
2
4-(Trifluoromethyl)phenylacetonitrile, 98%, Thermo Scientific™
CAS: 2338-75-2 Molecular Formula: C9H6F3N Molecular Weight (g/mol): 185.15 MDL Number: MFCD00001921 InChI Key: QNKOCFJZJWOXDE-UHFFFAOYSA-N Synonym: 4-trifluoromethyl phenylacetonitrile,2-4-trifluoromethyl phenyl acetonitrile,4-trifluoromethyl benzyl cyanide,p-trifluoromethylbenzyl cyanide,4-trifluoromethyl phenyl acetonitrile,4-trifluoromethyl-phenyl-acetonitrile,benzeneacetonitrile, 4-trifluoromethyl,4-trifluoromethylbenzyl cyanide,p-trifluoromethylphenylacetonitrile PubChem CID: 75359 IUPAC Name: 2-[4-(trifluoromethyl)phenyl]acetonitrile SMILES: FC(F)(F)C1=CC=C(CC#N)C=C1