accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOO

CAS RN 23405-15-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl benzoate
Synonyms: 2,5-dioxopyrrolidin-1-yl benzoate
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: BVUOEDOMUOJKOY-UHFFFAOYSA-N
SMILES: O=C(ON1C(=O)CCC1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results