accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3O

CAS RN 2345-28-0

IUPAC Name: pentadecan-2-one
Synonyms: 2-pentadecanone
Molecular Weight (g/mol): 226.4
Molecular Formula: C15H30O
InChi Key: CJPNOLIZCWDHJK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)