accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23519-77-9

IUPAC Name: zirconium(4+) tetrakis(propan-1-olate)
Synonyms: zirconium(4+) tetrakis(propan-1-olate)
Molecular Weight (g/mol): 327.58
Molecular Formula: C12H28O4Zr
InChi Key: XPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILES: [Zr+4].CCC[O-].CCC[O-].CCC[O-].CCC[O-]
Physical Form 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results