CAS RN 2353-33-5
5-Aza-2'-deoxycytidine, 98%
CAS: 2353-33-5 Molecular Formula: C9H13N3O4 Molecular Weight (g/mol): 227.22 MDL Number: MFCD00006547 InChI Key: CKTSBUTUHBMZGZ-SHYZEUOFSA-N Synonym: decitabine,5-aza-2'-deoxycytidine,dacogen,5-azadeoxycytidine,2'-deoxy-5-azacytidine,azadc,dezocitidine,5-aza-cdr,5-aza-dc,dacogen tn PubChem CID: 451668 ChEBI: CHEBI:50131 IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one SMILES: NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
5-Aza-2'-deoxycytidine, 98%
CAS: 2353-33-5 Molecular Formula: C9H13N3O4 Molecular Weight (g/mol): 227.22 MDL Number: MFCD00006547 InChI Key: CKTSBUTUHBMZGZ-SHYZEUOFSA-N Synonym: decitabine,5-aza-2'-deoxycytidine,dacogen,5-azadeoxycytidine,2'-deoxy-5-azacytidine,azadc,dezocitidine,5-aza-cdr,5-aza-dc,dacogen tn PubChem CID: 451668 ChEBI: CHEBI:50131 IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one SMILES: NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
5-Aza-2'-deoxycytidine, MP Biomedicals™
CAS: 2353-33-5 Molecular Formula: C9H13N3O4 Molecular Weight (g/mol): 227.22 MDL Number: MFCD00006547 InChI Key: CKTSBUTUHBMZGZ-SHYZEUOFSA-N Synonym: decitabine,5-aza-2'-deoxycytidine,dacogen,5-azadeoxycytidine,2'-deoxy-5-azacytidine,azadc,dezocitidine,5-aza-cdr,5-aza-dc,dacogen tn PubChem CID: 451668 ChEBI: CHEBI:50131 SMILES: NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
R&D Systems™ Decitabine
CAS: 2353-33-5 Molecular Formula: C9H13N3O4
Decitabine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Aza-2’-deoxy Cytidine, TRC
CAS: 2353-33-5 Molecular Formula: C8 H12 N4 O4 Molecular Weight (g/mol): 228.21 Synonym: 1,3,5-Triazin-2(1H)-one, 4-amino-1-(2-deoxy-β-D-erythro-pentofuranosyl)-,s-Triazin-2(1H)-one, 4-amino-1-(2-deoxy-β-D-erythro-pentofuranosyl)- (7CI,8CI),4-Amino-1-(2-deoxy-β-D-erythro-pentofuranosyl)-1,3,5-triazin-2(1H)-one,2-Desoxy-5-azacytidine,2'-Deoxy-5-azacytidine,4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one,5-Aza-2-deoxycytidine,5-Aza-2'-deoxycytidine,5-Azadeoxycytidine,DAC,Dacogen,Decitabine,NSC 127716,β-Decitabine IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one SMILES: NC1=NC(=O)N(C=N1)[C@H]2C[C@H](O)[C@@H](CO)O2