accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FSOOFFF

CAS RN 2358-41-0

IUPAC Name: 1-fluoro-2-trifluoromethanesulfonylbenzene
Synonyms: 1-fluoro-2-trifluoromethanesulfonylbenzene
Molecular Weight (g/mol): 228.16
Molecular Formula: C7H4F4O2S
InChi Key: NXZFXDLCZKKNCX-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results