accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNN

CAS RN 23593-75-1

IUPAC Name: 1-[(2-chlorophenyl)diphenylmethyl]-1H-imidazole
Synonyms: clotrimazole lotrimax gynix mycelex cream rimazole lotrimin lotion clotrimaderm cream mycelax otomax canesten 3-day therapy
Molecular Weight (g/mol): 344.84
Molecular Formula: C22H17ClN2
InChi Key: VNFPBHJOKIVQEB-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(N1C=CN=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Certifications/Compliance 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (9)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)