accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2361-27-5

NH2HNOS

CAS RN 2361-27-5

IUPAC Name: thiophene-2-carbohydrazide
Synonyms: thiophene-2-carbohydrazide
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: SOGBOGBTIKMGFS-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)