accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2362-62-1

CH3NH2S

CAS RN 2362-62-1

IUPAC Name: 4-methylbenzene-1-carbothioamide
Synonyms: 4-methylbenzenecarbothioamide
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: QXYZSNGZMDVLKN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results