CAS RN 236395-14-5
IUPAC Name:
(9S)-2-carbamoyl-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-9-yl acetate
Synonyms:
eslicarbazepine acetate
Molecular Weight (g/mol):
296.33
Molecular Formula:
C17H16N2O3
InChi Key:
QIALRBLEEWJACW-INIZCTEOSA-N
SMILES:
CC(=O)O[C@H]1CC2=CC=CC=C2N(C(N)=O)C2=CC=CC=C12
1
–
4
of
4
results
1
Eslicarbazepine Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Eslicarbazepine Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Eslicarbazepine Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Eslicarbazepine Acetate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.