accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NH2H3CH3CCH3

CAS RN 2380-36-1

IUPAC Name: 3,5-di-tert-butylaniline
Synonyms: 3,5-di-tert-butylaniline
Molecular Weight (g/mol): 205.35
Molecular Formula: C14H23N
InChi Key: MJKNHXCPGXUEDO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(N)=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results