accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2380-84-9

OHHN

CAS RN 2380-84-9

IUPAC Name: 1H-indol-7-ol
Synonyms: 1H-indol-7-ol
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: ORVPXPKEZLTMNW-UHFFFAOYSA-N
SMILES: OC1=C2NC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results