accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHN

CAS RN 2380-86-1

IUPAC Name: 1H-indol-6-ol
Synonyms: 1H-indol-6-ol
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: XAWPKHNOFIWWNZ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CNC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)