accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23806-24-8

CH3SOO

CAS RN 23806-24-8

IUPAC Name: 3-methylthiophene-2-carboxylate
Synonyms: 3-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 141.16
Molecular Formula: C6H5O2S
InChi Key: IFLKEBSJTZGCJG-UHFFFAOYSA-M
SMILES: CC1=C(SC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results