accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2382-96-9

IUPAC Name: 2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 2(3H)-benzoxazolethione
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: FLFWJIBUZQARMD-UHFFFAOYSA-N
SMILES: S=C1NC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)