accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23853-33-0

IUPAC Name: [(hydroxymethyl)sulfanyl](phenyl)methanone
Synonyms: [(hydroxymethyl)sulfanyl](phenyl)methanone
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: MNTZJQAAOYSCMM-UHFFFAOYSA-N
SMILES: OCSC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results