accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2386-57-4

NaCH3SOHOO

CAS RN 2386-57-4

IUPAC Name: methanesulfonic acid sodium
Synonyms: methanesulfonic acid sodium
Molecular Weight (g/mol): 119.09
Molecular Formula: CH4NaO3S
InChi Key: DWNWZFAKFWMPTI-UHFFFAOYSA-N
SMILES: [Na].CS(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results