accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CMgCl

CAS RN 2386-64-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 88.82
Molecular Formula: C2H5ClMg
InChi Key: YCCXQARVHOPWFJ-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].C[CH2-]
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)