accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NOH2NHNNO

CAS RN 2387-48-6

IUPAC Name: 2,6-diamino-5-nitroso-1,4-dihydropyrimidin-4-one
Synonyms: 2,6-diamino-5-nitroso-1H-pyrimidin-4-one
Molecular Weight (g/mol): 155.12
Molecular Formula: C4H5N5O2
InChi Key: HVMRLFSFHWCUCG-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C(N=O)=C(N)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results