accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 238742-86-4

IUPAC Name: 5-({[5-(trifluoromethyl)pyridin-2-yl]sulfanyl}methyl)furan-2-carboxylic acid
Synonyms: 5-({[5-(trifluoromethyl)pyridin-2-yl]sulfanyl}methyl)furan-2-carboxylic acid
Molecular Weight (g/mol): 303.26
Molecular Formula: C12H8F3NO3S
InChi Key: MYDIGMNTURCKBI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CSC2=NC=C(C=C2)C(F)(F)F)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results