accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNI

CAS RN 23889-85-2

IUPAC Name: 4-iodo-1-phenyl-1H-pyrazole
Synonyms: 4-iodo-1-phenylpyrazole
Molecular Weight (g/mol): 270.07
Molecular Formula: C9H7IN2
InChi Key: PYJVDCYRDQUUKO-UHFFFAOYSA-N
SMILES: IC1=CN(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results