accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23894-12-4

IUPAC Name: 6-aminonaphthalen-1-ol
Synonyms: 6-aminonaphthalen-1-ol
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: QYFYIOWLBSPSDM-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(O)=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results