accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH3CNH3CSCH3NH3C

CAS RN 2390-54-7

IUPAC Name: 2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
Synonyms: thioflavin
Molecular Weight (g/mol): 318.86
Molecular Formula: C17H19ClN2S
InChi Key: JADVWWSKYZXRGX-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1
Color 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)