accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH3CNH3CCH3CH3

CAS RN 2390-68-3

IUPAC Name: bis(decyl)dimethylazanium bromide
Synonyms: didecyl-dimethyl-ammonium bromide
Molecular Weight (g/mol): 406.54
Molecular Formula: C22H48BrN
InChi Key: UMGXUWVIJIQANV-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCC[N+](C)(C)CCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)