accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 239135-48-9

IUPAC Name: 2-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: MQZWBTRGSDOAIO-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results