accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 239135-52-5

IUPAC Name: 2-[5-fluoro-2-(trifluoromethyl)phenyl]acetic acid
Synonyms: [5-fluoro-2-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: GWWZRGDMAFXULO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(C=CC(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results