accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONH2

CAS RN 2393-23-9

IUPAC Name: 1-(4-methoxyphenyl)methanamine
Synonyms: benzenemethanamine, 4-methoxy-
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: IDPURXSQCKYKIJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)