accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NClFF

CAS RN 239463-89-9

IUPAC Name: 4-chloro-6,8-difluoroquinoline
Synonyms: 4-chloro-6,8-difluoroquinoline
Molecular Weight (g/mol): 199.58
Molecular Formula: C9H4ClF2N
InChi Key: NBYDBLFWTPXYFH-UHFFFAOYSA-N
SMILES: FC1=CC2=C(Cl)C=CN=C2C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results