accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOFFFF(E/Z)

CAS RN 239463-90-2

IUPAC Name: 3-[4-fluoro-3-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[4-fluoro-3-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 234.15
Molecular Formula: C10H6F4O2
InChi Key: DESJUJUEXMNIGB-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(=C(F)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results