accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HCCH

CAS RN 2396-63-6

IUPAC Name: hepta-1,6-diyne
Synonyms: hepta-1,6-diyne
Molecular Weight (g/mol): 92.14
Molecular Formula: C7H8
InChi Key: RSPZSDWVQWRAEF-UHFFFAOYSA-N
SMILES: C#CCCCC#C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)