accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOONHOCH

CAS RN 2398-81-4

IUPAC Name: 5-carboxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Synonyms: 5-carboxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Molecular Weight (g/mol): 140.12
Molecular Formula: C6H6NO3
InChi Key: GDILJONVBZVAFH-UHFFFAOYNA-N
SMILES: OC(=O)C1=C[NH++]([O-])[CH-]C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results