accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23995-88-2

IUPAC Name: 1-(1-methylpiperidin-4-yl)piperazine
Synonyms: 1-(1-methylpiperidin-4-yl)piperazine
Molecular Weight (g/mol): 183.3
Molecular Formula: C10H21N3
InChi Key: OHUMKYGINIODOY-UHFFFAOYSA-N
SMILES: CN1CCC(CC1)N1CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)